5-Bromo-3-methyl-N-(4-pyridinylmethyl)-2-pyridinamine
Supplier: Matrix Scientific
053467-500MG
053467-2.5G
101903-754EA
364.62
USD
101903-754
101903-756
5-Bromo-3-methyl-N-(4-pyridinylmethyl)-2-pyridinamine
5-Bromo-3-methyl-N-(4-pyridinylmethyl)-2-pyridinamine
Hazard: Irritant
Formula:
C₁₂H₁₂BrN₃ |
MDL Number:
MFCD13562802 CAS Number: 1219976-58-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...