2-Chloro-N-(4-pyridinylmethyl)nicotinamide

Supplier: Matrix Scientific

053466-500MG 053466-2.5G
101903-750EA 364.62 USD
101903-750 101903-752
2-Chloro-N-(4-pyridinylmethyl)nicotinamide
2-Chloro-N-(4-pyridinylmethyl)nicotinamide

Hazard: Irritant

Formula: C₁₂H₁₀ClN₃O
MDL Number: MFCD03382940
CAS Number: 224048-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR