N2-(4-Pyridinylmethyl)-2,3-pyridinediamine

Supplier: Matrix Scientific

053465-500MG 053465-2.5G
101903-746EA 364.62 USD
101903-746 101903-748
N2-(4-Pyridinylmethyl)-2,3-pyridinediamine
N2-(4-Pyridinylmethyl)-2,3-pyridinediamine

Hazard: Irritant

Formula: C₁₁H₁₂N₄
MDL Number: MFCD11187333
CAS Number: 1040316-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR