2-[(4-Pyridinylmethyl)amino]isonicotinic acid

Supplier: Matrix Scientific

053463-500MG 053463-2.5G
101903-738EA 384.62 USD
101903-738 101903-740
2-[(4-Pyridinylmethyl)amino]isonicotinic acid
2-[(4-Pyridinylmethyl)amino]isonicotinic acid

Hazard: Irritant

Formula: C₁₂H₁₁N₃O₂
MDL Number: MFCD11116740
CAS Number: 1036467-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR