6-Chloro-N-(4-pyridinylmethyl)-2-pyrazinamine

Supplier: Matrix Scientific

053462-500MG 053462-2.5G
101903-734EA 149.23 USD
101903-734 101903-736
6-Chloro-N-(4-pyridinylmethyl)-2-pyrazinamine
6-Chloro-N-(4-pyridinylmethyl)-2-pyrazinamine

Hazard: Irritant

Formula: C₁₀H₉ClN₄
MDL Number: MFCD13562801
CAS Number: 1220030-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR