3-Chloro-N-(4-pyridinylmethyl)-5-(trifluoromethyl)-2-pyridinamine

Supplier: Matrix Scientific

053461-500MG 053461-2.5G
101903-730EA 364.62 USD
101903-730 101903-732
3-Chloro-N-(4-pyridinylmethyl)-5-(trifluoromethyl)-2-pyridinamine
3-Chloro-N-(4-pyridinylmethyl)-5-(trifluoromethyl)-2-pyridinamine

Hazard: Irritant

Formula: C₁₂H₉ClF₃N₃
MDL Number: MFCD12620677
CAS Number: 1040046-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR