6-Chloro-N-(4-pyridinylmethyl)-4-pyrimidinamine
Supplier: Matrix Scientific
053460-500MG
053460-2.5G
101903-726EA
364.62
USD
101903-726
101903-728
6-Chloro-N-(4-pyridinylmethyl)-4-pyrimidinamine
6-Chloro-N-(4-pyridinylmethyl)-4-pyrimidinamine
Hazard: Irritant
Formula:
C₁₀H₉ClN₄ |
MDL Number:
MFCD11655643 CAS Number: 1086386-59-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...