5-Bromo-4-methyl-N-(4-pyridinylmethyl)-2-pyridinamine

Supplier: Matrix Scientific

053459-500MG 053459-2.5G
101903-722EA 364.62 USD
101903-722 101903-724
5-Bromo-4-methyl-N-(4-pyridinylmethyl)-2-pyridinamine
5-Bromo-4-methyl-N-(4-pyridinylmethyl)-2-pyridinamine

Hazard: Irritant

Formula: C₁₂H₁₂BrN₃
MDL Number: MFCD13562800
CAS Number: 1220036-42-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR