3-Nitro-4-[(4-pyridinylmethyl)amino]benzoic acid

Supplier: Matrix Scientific

053458-500MG 053458-2.5G
101903-718EA 364.62 USD
101903-718 101903-720
3-Nitro-4-[(4-pyridinylmethyl)amino]benzoic acid
3-Nitro-4-[(4-pyridinylmethyl)amino]benzoic acid

Hazard: Irritant

Formula: C₁₃H₁₁N₃O₄
MDL Number: MFCD13355474
CAS Number: 1041533-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR