3-Amino-4-chloro-N-(2-pyridinylmethyl)benzenesulfonamide

Supplier: Matrix Scientific

053456-500MG 053456-2.5G
101903-710EA 364.62 USD
101903-710 101903-712
3-Amino-4-chloro-N-(2-pyridinylmethyl)benzenesulfonamide
3-Amino-4-chloro-N-(2-pyridinylmethyl)benzenesulfonamide

Hazard: Irritant

Formula: C₁₂H₁₂ClN₃O₂S
MDL Number: MFCD12440589
CAS Number: 1036494-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR