N-(2-Pyridinylmethyl)-4-piperidinecarboxamide hydrochloride
Supplier: Matrix Scientific
053454-500MG
053454-2.5G
101903-702EA
364.62
USD
101903-702
101903-704
N-(2-Pyridinylmethyl)-4-piperidinecarboxamide hydrochloride
N-(2-Pyridinylmethyl)-4-piperidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₈ClN₃O |
MDL Number:
MFCD13562797 CAS Number: 1219957-82-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...