N-(2-Pyridinylmethyl)-3-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

053453-500MG 053453-2.5G
101903-698EA 364.62 USD
101903-698 101903-700
N-(2-Pyridinylmethyl)-3-piperidinecarboxamide hydrochloride
N-(2-Pyridinylmethyl)-3-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₈ClN₃O
MDL Number: MFCD13562796
CAS Number: 1220036-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR