N-(2-Pyridinylmethyl)-2-pyrrolidinecarboxamide hydrochloride

Supplier: Matrix Scientific

053451-500MG 053451-2.5G
101903-690EA 364.62 USD
101903-690 101903-692
N-(2-Pyridinylmethyl)-2-pyrrolidinecarboxamide hydrochloride
N-(2-Pyridinylmethyl)-2-pyrrolidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₆ClN₃O
MDL Number: MFCD13562794
CAS Number: 1236261-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR