2-Amino-3-phenyl-N-(2-pyridinylmethyl)propanamide hydrochloride
Supplier: Matrix Scientific
053450-500MG
053450-2.5G
101903-686EA
364.62
USD
101903-686
101903-688
2-Amino-3-phenyl-N-(2-pyridinylmethyl)propanamide hydrochloride
2-Amino-3-phenyl-N-(2-pyridinylmethyl)propanamide hydrochloride
Hazard: Irritant
Formula:
C₁₅H₁₈ClN₃O |
MDL Number:
MFCD13562793 CAS Number: 1246172-53-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...