2-Amino-3-methyl-N-(2-pyridinylmethyl)butanamide hydrochloride

Supplier: Matrix Scientific

053448-500MG 053448-2.5G
101903-678EA 364.62 USD
101903-678 101903-680
2-Amino-3-methyl-N-(2-pyridinylmethyl)butanamide hydrochloride
2-Amino-3-methyl-N-(2-pyridinylmethyl)butanamide hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₈ClN₃O
MDL Number: MFCD13562791
CAS Number: 1236261-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR