3-Amino-N-(2-pyridinylmethyl)propanamide hydrochloride

Supplier: Matrix Scientific

053447-500MG 053447-2.5G
101903-674EA 149.23 USD
101903-674 101903-676
3-Amino-N-(2-pyridinylmethyl)propanamide hydrochloride
3-Amino-N-(2-pyridinylmethyl)propanamide hydrochloride

Hazard: Irritant

Formula: C₉H₁₄ClN₃O
MDL Number: MFCD13562790
CAS Number: 1220035-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR