2-Amino-N-(2-pyridinylmethyl)propanamide hydrochloride

Supplier: Matrix Scientific

053445-500MG 053445-2.5G
101903-666EA 364.62 USD
101903-666 101903-668
2-Amino-N-(2-pyridinylmethyl)propanamide hydrochloride
2-Amino-N-(2-pyridinylmethyl)propanamide hydrochloride

Hazard: Irritant

Formula: C₉H₁₄ClN₃O
MDL Number: MFCD13562788
CAS Number: 1236267-72-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR