2-Amino-N-(2-pyridinylmethyl)acetamide hydrochloride
Supplier: Matrix Scientific
053444-500MG
053444-2.5G
101903-662EA
364.62
USD
101903-662
101903-664
2-Amino-N-(2-pyridinylmethyl)acetamide hydrochloride
2-Amino-N-(2-pyridinylmethyl)acetamide hydrochloride
Hazard: Irritant
Formula:
C₈H₁₂ClN₃O |
MDL Number:
MFCD13562787 CAS Number: 1220020-36-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...