6-Chloro-N-(2-pyridinylmethyl)-2-pyridinamine

Supplier: Matrix Scientific

053443-500MG 053443-2.5G
101903-658EA 364.62 USD
101903-658 101903-660
6-Chloro-N-(2-pyridinylmethyl)-2-pyridinamine
6-Chloro-N-(2-pyridinylmethyl)-2-pyridinamine

Hazard: Irritant

Formula: C₁₁H₁₀ClN₃
MDL Number: MFCD13562786
CAS Number: 142889-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR