5-Bromo-N-(2-pyridinylmethyl)-2-pyridinamine

Supplier: Matrix Scientific

053442-500MG 053442-2.5G
101903-654EA 364.62 USD
101903-654 101903-656
5-Bromo-N-(2-pyridinylmethyl)-2-pyridinamine
5-Bromo-N-(2-pyridinylmethyl)-2-pyridinamine

Hazard: Irritant

Formula: C₁₁H₁₀BrN₃
MDL Number: MFCD11122322
CAS Number: 1041542-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR