5-Bromo-3-methyl-N-(2-pyridinylmethyl)-2-pyridinamine

Supplier: Matrix Scientific

053441-500MG 053441-2.5G
101903-650EA 364.62 USD
101903-650 101903-652
5-Bromo-3-methyl-N-(2-pyridinylmethyl)-2-pyridinamine
5-Bromo-3-methyl-N-(2-pyridinylmethyl)-2-pyridinamine

Hazard: Irritant

Formula: C₁₂H₁₂BrN₃
MDL Number: MFCD13562785
CAS Number: 1220035-38-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR