N2-(2-Pyridinylmethyl)-2,3-pyridinediamine
Supplier: Matrix Scientific
053439-500MG
053439-2.5G
101903-642EA
364.62
USD
101903-642
101903-644
N2-(2-Pyridinylmethyl)-2,3-pyridinediamine
N2-(2-Pyridinylmethyl)-2,3-pyridinediamine
Hazard: Irritant
Formula:
C₁₁H₁₂N₄ |
MDL Number:
MFCD11101079 CAS Number: 99138-92-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...