2-[(2-Pyridinylmethyl)amino]nicotinic acid
Supplier: Matrix Scientific
053438-500MG
053438-2.5G
101903-638EA
364.62
USD
101903-638
101903-640
2-[(2-Pyridinylmethyl)amino]nicotinic acid
2-[(2-Pyridinylmethyl)amino]nicotinic acid
Hazard: Irritant
Formula:
C₁₂H₁₁N₃O₂ |
MDL Number:
MFCD11132164 CAS Number: 1019358-70-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...