2-[(2-Pyridinylmethyl)amino]isonicotinic acid
Supplier: Matrix Scientific
053437-500MG
053437-2.5G
101903-634EA
364.62
USD
101903-634
101903-636
2-[(2-Pyridinylmethyl)amino]isonicotinic acid
2-[(2-Pyridinylmethyl)amino]isonicotinic acid
Hazard: Irritant
Formula:
C₁₂H₁₁N₃O₂ MW: 229.24 g/mol |
MDL Number:
MFCD11132162 CAS Number: 1019322-73-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...