6-Chloro-N-(2-pyridinylmethyl)-2-pyrazinamine
Supplier: Matrix Scientific
053436-500MG
053436-2.5G
101903-630EA
364.62
USD
101903-630
101903-632
6-Chloro-N-(2-pyridinylmethyl)-2-pyrazinamine
6-Chloro-N-(2-pyridinylmethyl)-2-pyrazinamine
Hazard: Irritant
Formula:
C₁₀H₉ClN₄ |
MDL Number:
MFCD13562784 CAS Number: 874143-67-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...