5-Bromo-4-methyl-N-(2-pyridinylmethyl)-2-pyridinamine

Supplier: Matrix Scientific

053433-500MG 053433-2.5G
101903-618EA 364.62 USD
101903-618 101903-620
5-Bromo-4-methyl-N-(2-pyridinylmethyl)-2-pyridinamine
5-Bromo-4-methyl-N-(2-pyridinylmethyl)-2-pyridinamine

Hazard: Irritant

Formula: C₁₂H₁₂BrN₃
MDL Number: MFCD13562782
CAS Number: 1220035-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR