3-Nitro-4-[(2-pyridinylmethyl)amino]benzoic acid

Supplier: Matrix Scientific

053432-500MG 053432-2.5G
101903-614EA 364.62 USD
101903-614 101903-616
3-Nitro-4-[(2-pyridinylmethyl)amino]benzoic acid
3-Nitro-4-[(2-pyridinylmethyl)amino]benzoic acid

Hazard: Irritant

Formula: C₁₃H₁₁N₃O₄
MDL Number: MFCD02937561
CAS Number: 440347-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR