Ethyl-3-amino-4-[(2-pyridinylmethyl)amino]benzoate

Supplier: Matrix Scientific

053431-500MG 053431-2.5G
101903-610EA 364.62 USD
101903-610 101903-612
Ethyl-3-amino-4-[(2-pyridinylmethyl)amino]benzoate
Ethyl-3-amino-4-[(2-pyridinylmethyl)amino]benzoate

Hazard: Irritant

Formula: C₁₅H₁₇N₃O₂
MDL Number: MFCD13562781
CAS Number: 1220035-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR