N-(3-Pyridinylmethyl)-3-piperidinecarboxamide hydrochloride
Supplier: Matrix Scientific
053427-500MG
053427-2.5G
101903-594EA
364.62
USD
101903-594
101903-596
N-(3-Pyridinylmethyl)-3-piperidinecarboxamide hydrochloride
N-(3-Pyridinylmethyl)-3-piperidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₈ClN₃O |
MDL Number:
MFCD13562778 CAS Number: 1220035-37-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...