N-(3-Pyridinylmethyl)-2-piperidinecarboxamide hydrochloride
Supplier: Matrix Scientific
053426-500MG
053426-2.5G
101903-590EA
364.62
USD
101903-590
101903-592
N-(3-Pyridinylmethyl)-2-piperidinecarboxamide hydrochloride
N-(3-Pyridinylmethyl)-2-piperidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₈ClN₃O |
MDL Number:
MFCD13562777 CAS Number: 1236255-22-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...