N-(3-Pyridinylmethyl)-2-pyrrolidinecarboxamide hydrochloride
Supplier: Matrix Scientific
053425-500MG
053425-2.5G
101903-586EA
364.62
USD
101903-586
101903-588
N-(3-Pyridinylmethyl)-2-pyrrolidinecarboxamide hydrochloride
N-(3-Pyridinylmethyl)-2-pyrrolidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₆ClN₃O MW: 241.72 g/mol |
MDL Number:
MFCD13562776 CAS Number: 1246172-90-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...