2-Amino-3-phenyl-N-(3-pyridinylmethyl)propanamide hydrochloride
Supplier: Matrix Scientific
053424-500MG
053424-2.5G
101903-582EA
364.62
USD
101903-582
101903-584
2-Amino-3-phenyl-N-(3-pyridinylmethyl)propanamide hydrochloride
2-Amino-3-phenyl-N-(3-pyridinylmethyl)propanamide hydrochloride
Hazard: Irritant
Formula:
C₁₅H₁₈ClN₃O |
MDL Number:
MFCD13562775 CAS Number: 1246172-57-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...