2-Amino-3-methyl-N-(3-pyridinylmethyl)butanamide hydrochloride

Supplier: Matrix Scientific

053422-500MG 053422-2.5G
101903-574EA 364.62 USD
101903-574 101903-576
2-Amino-3-methyl-N-(3-pyridinylmethyl)butanamide hydrochloride
2-Amino-3-methyl-N-(3-pyridinylmethyl)butanamide hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₈ClN₃O
MDL Number: MFCD13562773
CAS Number: 1236257-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR