3-Amino-N-(3-pyridinylmethyl)propanamide hydrochloride
Supplier: Matrix Scientific
053421-500MG
053421-2.5G
101903-570EA
364.62
USD
101903-570
101903-572
3-Amino-N-(3-pyridinylmethyl)propanamide hydrochloride
3-Amino-N-(3-pyridinylmethyl)propanamide hydrochloride
Hazard: Irritant
Formula:
C₉H₁₄ClN₃O |
MDL Number:
MFCD13195875 CAS Number: 1208947-75-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...