2-(Methylamino)-N-(3-pyridinylmethyl)acetamide hydrochloride

Supplier: Matrix Scientific

053420-500MG 053420-2.5G
101903-566EA 364.62 USD
101903-566 101903-568
2-(Methylamino)-N-(3-pyridinylmethyl)acetamide hydrochloride
2-(Methylamino)-N-(3-pyridinylmethyl)acetamide hydrochloride

Hazard: Irritant

Formula: C₉H₁₄ClN₃O
MDL Number: MFCD13562772
CAS Number: 1220038-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR