2-Amino-N-(3-pyridinylmethyl)propanamide hydrochloride
Supplier: Matrix Scientific
053419-500MG
053419-2.5G
101903-562EA
364.62
USD
101903-562
101903-564
2-Amino-N-(3-pyridinylmethyl)propanamide hydrochloride
2-Amino-N-(3-pyridinylmethyl)propanamide hydrochloride
Hazard: Irritant
Formula:
C₉H₁₄ClN₃O |
MDL Number:
MFCD13562771 CAS Number: 1236261-24-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...