2-Amino-N-(3-pyridinylmethyl)acetamide hydrochloride
Supplier: Matrix Scientific
053418-500MG
053418-2.5G
101903-558EA
166.15
USD
101903-558
101903-560
2-Amino-N-(3-pyridinylmethyl)acetamide hydrochloride
2-Amino-N-(3-pyridinylmethyl)acetamide hydrochloride
Hazard: Irritant
Formula:
C₈H₁₂ClN₃O |
MDL Number:
MFCD13562770 CAS Number: 1220035-35-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...