6-Chloro-N-(3-pyridinylmethyl)-2-pyridinamine
Supplier: Matrix Scientific
053417-500MG
053417-2.5G
101903-554EA
461.54
USD
101903-554
101903-556
6-Chloro-N-(3-pyridinylmethyl)-2-pyridinamine
6-Chloro-N-(3-pyridinylmethyl)-2-pyridinamine
Hazard: Irritant
Formula:
C₁₁H₁₀ClN₃ |
MDL Number:
MFCD13562769 CAS Number: 1220038-38-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...