5-Bromo-N-(3-pyridinylmethyl)-2-pyridinamine

Supplier: Matrix Scientific

053416-500MG 053416-2.5G
101903-550EA 364.62 USD
101903-550 101903-552
5-Bromo-N-(3-pyridinylmethyl)-2-pyridinamine
5-Bromo-N-(3-pyridinylmethyl)-2-pyridinamine

Hazard: Irritant

Formula: C₁₁H₁₀BrN₃
MDL Number: MFCD11117959
CAS Number: 1036505-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR