5-Bromo-3-methyl-N-(3-pyridinylmethyl)-2-pyridinamine

Supplier: Matrix Scientific

053415-500MG 053415-2.5G
101903-546EA 364.62 USD
101903-546 101903-548
5-Bromo-3-methyl-N-(3-pyridinylmethyl)-2-pyridinamine
5-Bromo-3-methyl-N-(3-pyridinylmethyl)-2-pyridinamine

Hazard: Irritant

Formula: C₁₂H₁₂BrN₃
MDL Number: MFCD13562768
CAS Number: 1219976-57-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR