N2-(3-Pyridinylmethyl)-2,3-pyridinediamine
Supplier: Matrix Scientific
053413-500MG
053413-2.5G
101903-538EA
364.62
USD
101903-538
101903-540
N2-(3-Pyridinylmethyl)-2,3-pyridinediamine
N2-(3-Pyridinylmethyl)-2,3-pyridinediamine
Hazard: Irritant
Formula:
C₁₁H₁₂N₄ |
MDL Number:
MFCD11101078 CAS Number: 1040064-25-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...