2-[(3-Pyridinylmethyl)amino]nicotinic acid
Supplier: Matrix Scientific
053412-500MG
053412-2.5G
101903-534EA
364.62
USD
101903-534
101903-536
2-[(3-Pyridinylmethyl)amino]nicotinic acid
2-[(3-Pyridinylmethyl)amino]nicotinic acid
Hazard: Irritant
Formula:
C₁₂H₁₁N₃O₂ |
MDL Number:
MFCD03114541 CAS Number: 460363-72-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...