2-[(3-Pyridinylmethyl)amino]isonicotinic acid
Supplier: Matrix Scientific
053411-500MG
053411-2.5G
101903-530EA
364.62
USD
101903-530
101903-532
2-[(3-Pyridinylmethyl)amino]isonicotinic acid
2-[(3-Pyridinylmethyl)amino]isonicotinic acid
Hazard: Irritant
Formula:
C₁₂H₁₁N₃O₂ |
MDL Number:
MFCD11117821 CAS Number: 1036599-81-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...