3-Nitro-4-[(3-pyridinylmethyl)amino]benzoic acid
Supplier: Matrix Scientific
053406-500MG
053406-2.5G
101903-510EA
364.62
USD
101903-510
101903-512
3-Nitro-4-[(3-pyridinylmethyl)amino]benzoic acid
3-Nitro-4-[(3-pyridinylmethyl)amino]benzoic acid
Hazard: Irritant
Formula:
C₁₃H₁₁N₃O₄ |
MDL Number:
MFCD02221224 CAS Number: 384855-46-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...