Ethyl-3-amino-4-[(3-pyridinylmethyl)amino]benzoate

Supplier: Matrix Scientific

053405-500MG 053405-2.5G
101903-506EA 364.62 USD
101903-506 101903-508
Ethyl-3-amino-4-[(3-pyridinylmethyl)amino]benzoate
Ethyl-3-amino-4-[(3-pyridinylmethyl)amino]benzoate

Hazard: Irritant

Formula: C₁₅H₁₇N₃O₂
MDL Number: MFCD13562764
CAS Number: 1220035-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR