N-Benzyl-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride

Supplier: Matrix Scientific

053403-500MG 053403-2.5G
101903-498EA 364.62 USD
101903-498 101903-500
N-Benzyl-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride
N-Benzyl-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride

Hazard: Irritant

Formula: C₁₄H₁₇ClN₄O
MW: 292.77 g/mol
MDL Number: MFCD13562763
CAS Number: 1220038-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR