N-Benzyl-4-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

053402-500MG 053402-2.5G
101903-494EA 364.62 USD
101903-494 101903-496
N-Benzyl-4-piperidinecarboxamide hydrochloride
N-Benzyl-4-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉ClN₂O
MDL Number: MFCD07329874
CAS Number: 101264-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR