N-Benzyl-2-piperidinecarboxamide hydrochloride
Supplier: Matrix Scientific
053400-500MG
053400-2.5G
101903-486EA
364.62
USD
101903-486
101903-488
N-Benzyl-2-piperidinecarboxamide hydrochloride
N-Benzyl-2-piperidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₁₃H₁₉ClN₂O |
MDL Number:
MFCD13562761 CAS Number: 205993-54-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...