N-Benzyl-2-pyrrolidinecarboxamide hydrochloride
Supplier: Matrix Scientific
Synonyms:
N-Benzylpyrrolidine-2-carboxamide hydrochloride
053399-500MG
053399-2.5G
101903-482EA
364.62
USD
101903-482
101903-484
N-Benzyl-2-pyrrolidinecarboxamide hydrochloride
N-Benzyl-2-pyrrolidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₇ClN₂O MW: 240.74 g/mol |
MDL Number:
MFCD13562760 CAS Number: 1236255-21-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...