2-Amino-N-benzyl-3-phenylpropanamide hydrochloride
Supplier: Matrix Scientific
053398-500MG
053398-2.5G
101903-478EA
364.62
USD
101903-478
101903-480
2-Amino-N-benzyl-3-phenylpropanamide hydrochloride
2-Amino-N-benzyl-3-phenylpropanamide hydrochloride
Hazard: Irritant
Formula:
C₁₆H₁₉ClN₂O MW: 290.8 g/mol |
MDL Number:
MFCD13562759 CAS Number: 34582-43-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...